ENA321764866 Enamine

3-[2-(propan-2-yl)-1H-imidazol-1-yl]propanoic acid hydrochloride

  • MDL number MFCD09997682
  • Cas Number : 1052526-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76295

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76295
Availability details for
Quantity
  • Availability
  • Add to Cart