ENA321764870 Enamine

2-amino-1-(3-methoxyphenyl)ethan-1-ol hydrochloride

  • MDL number MFCD05861640
  • Cas Number : 53517-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76961
Availability details for
Quantity
  • Availability
  • Add to Cart