ENA321764871 Enamine

3-cyclopropylimidazolidine-2,4-dione

  • MDL number MFCD16665293
  • Cas Number : 69627-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76962

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76962
Availability details for
Quantity
  • Availability
  • Add to Cart