ENA321764874 Enamine

2-(4-cyclopentanecarbonylpiperazin-1-yl)ethan-1-amine

  • MDL number MFCD11620415
  • Cas Number : 1094880-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76048
Availability details for
Quantity
  • Availability
  • Add to Cart