ENA321764877 Enamine

5-oxo-1-(3-sulfamoylphenyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD09812624
  • Cas Number : 63674-70-4
  • Molecular Weight : 284.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-76968

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76968
Availability details for
Quantity
  • Availability
  • Add to Cart