ENA321764879 Enamine

N-(4-hydroxyphenyl)-2-methoxyacetamide

  • MDL number MFCD05155980
  • Cas Number : 723757-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76300
Availability details for
Quantity
  • Availability
  • Add to Cart