ENA321764880 Enamine

4-[5-(bromomethyl)-4,5-dihydro-1,3-thiazol-2-yl]morpholine hydrobromide

  • MDL number MFCD00781261
  • Cas Number : 92504-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Product Link https://www.enaminestore.com/catalog/EN300-76301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76301
Availability details for
Quantity
  • Availability
  • Add to Cart