ENA321764881 Enamine

3-amino-1-[4-(trifluoromethyl)phenyl]pyrrolidin-2-one

  • MDL number MFCD16202975
  • Cas Number : 1270609-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76973
Availability details for
Quantity
  • Availability
  • Add to Cart