ENA321764894 Enamine

8,9-dimethyl-2,3,4,5-tetrahydro-1-benzoxepin-5-one

  • MDL number MFCD11104774
  • Cas Number : 141106-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76992
Availability details for
Quantity
  • Availability
  • Add to Cart