ENA321764898 Enamine

(butan-2-yl)(1-{[1,2,4]triazolo[4,3-a]pyridin-3-yl}ethyl)amine

  • MDL number MFCD12119935
  • Cas Number : 1156212-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76311
Availability details for
Quantity
  • Availability
  • Add to Cart