ENA321764901 Enamine

3-(3-phenoxyphenyl)-1,2,4-thiadiazol-5-amine

  • MDL number MFCD12682772
  • Cas Number : 1153983-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76999

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76999
Availability details for
Quantity
  • Availability
  • Add to Cart