ENA321764905 Enamine

2-chloro-4-methoxy-6-methylpyrimidine

  • MDL number MFCD08689881
  • Cas Number : 22536-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35...37
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77003

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77003
Availability details for
Quantity
  • Availability
  • Add to Cart