ENA321764907 Enamine

3-methyl-octahydropyrrolo[1,2-a]piperazine-1,4-dione

  • MDL number MFCD16991131
  • Cas Number : 65556-33-4
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-76315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76315
Availability details for
Quantity
  • Availability
  • Add to Cart