ENA321764908 Enamine

2-[(3-methyl-1,2,4-oxadiazol-5-yl)methoxy]acetic acid

  • MDL number MFCD14620962
  • Cas Number : 1083424-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77006
Availability details for
Quantity
  • Availability
  • Add to Cart