ENA321764925 Enamine

2,4-dimethylcyclohexan-1-amine

  • MDL number MFCD00130040
  • Cas Number : 28294-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76052

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76052
Availability details for
Quantity
  • Availability
  • Add to Cart