ENA321764929 Enamine

1-(1,4-dioxan-2-ylmethyl)piperazine

  • MDL number MFCD12779495
  • Cas Number : 329217-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76361

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76361
Availability details for
Quantity
  • Availability
  • Add to Cart