ENA321764932 Enamine

2-(phenylformamido)-3-(pyridin-3-yl)propanoic acid

  • MDL number MFCD18838781
  • Cas Number : 33560-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76372
Availability details for
Quantity
  • Availability
  • Add to Cart