ENA321764934 Enamine

4-(aminomethyl)oxan-4-amine

  • MDL number MFCD08234566
  • Cas Number : 50289-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76374

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76374
Availability details for
Quantity
  • Availability
  • Add to Cart