ENA321764941 Enamine

pentane-2-sulfonamide

  • MDL number MFCD01709292
  • Cas Number : 17854-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48...50
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77053

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77053
Availability details for
Quantity
  • Availability
  • Add to Cart