ENA321764942 Enamine

3-methoxy-5-methylthiophene-2-carboxylic acid

  • MDL number MFCD17012773
  • Cas Number : 83261-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77068
Availability details for
Quantity
  • Availability
  • Add to Cart