ENA321764944 Enamine

3-(2,4-difluorophenyl)-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11180206
  • Cas Number : 1152508-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77071
Availability details for
Quantity
  • Availability
  • Add to Cart