ENA321764948 Enamine

3-amino-3-cyclohexylpropan-1-ol

  • MDL number MFCD03412684
  • Cas Number : 109647-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77076

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77076
Availability details for
Quantity
  • Availability
  • Add to Cart