ENA321764955 Enamine

4-fluoro-2-(2,2,2-trifluoroethoxy)aniline

  • MDL number MFCD09738718
  • Cas Number : 186386-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77084

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77084
Availability details for
Quantity
  • Availability
  • Add to Cart