ENA321764970 Enamine

4-(piperidin-1-yl)pyridin-3-amine

  • MDL number MFCD10691294
  • Cas Number : 52311-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76385
Availability details for
Quantity
  • Availability
  • Add to Cart