ENA321764974 Enamine

methyl 3-[(2-methoxyethyl)amino]propanoate

  • MDL number MFCD10687305
  • Cas Number : 55694-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76397

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76397
Availability details for
Quantity
  • Availability
  • Add to Cart