ENA321764975 Enamine

3,4,5-trimethyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD00266483
  • Cas Number : 17106-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80...82
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76399

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-76399
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart