ENA321764978 Enamine

5-amino-1-(2-methylphenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD03934850
  • Cas Number : 14678-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76411

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76411
Availability details for
Quantity
  • Availability
  • Add to Cart