ENA321764980 Enamine

3-(propan-2-yl)-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD05668695
  • Cas Number : 14633-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74...76
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77144

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-77144
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart