ENA321764981 Enamine

1-(piperazin-1-yl)cyclopentane-1-carboxylic acid

  • MDL number MFCD12818024
  • Cas Number : 1183507-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76416

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76416
Availability details for
Quantity
  • Availability
  • Add to Cart