ENA321764983 Enamine

(3-methyloxolan-3-yl)methanamine

  • MDL number MFCD16856414
  • Cas Number : 864814-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76418

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-76418
pictograms GHS02,GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart