ENA321764987 Enamine

2-[4-(1-hydroxyethyl)phenoxy]-N,N-dimethylacetamide

  • MDL number MFCD08442881
  • Cas Number : 926242-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77153

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77153
Availability details for
Quantity
  • Availability
  • Add to Cart