ENA321764988 Enamine

methyl 2-(5-nitrothiophen-2-yl)acetate

  • MDL number MFCD12172986
  • Cas Number : 1154278-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77154
Availability details for
Quantity
  • Availability
  • Add to Cart