ENA321764998 Enamine

(5-bromo-2-{methyl[2-(pyridin-2-yl)ethyl]amino}pyridin-3-yl)methanol

  • MDL number MFCD15503824
  • Cas Number : 1306607-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77167

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77167
Availability details for
Quantity
  • Availability
  • Add to Cart