ENA321765001 Enamine

5-hydroxypentanenitrile

  • MDL number MFCD09746052
  • Cas Number : 2427-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77170
Availability details for
Quantity
  • Availability
  • Add to Cart