ENA321765009 Enamine

4-[(5-ethyl-1,2,4-oxadiazol-3-yl)methoxy]piperidine

  • MDL number MFCD14679482
  • Cas Number : 1251082-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77180

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77180
Availability details for
Quantity
  • Availability
  • Add to Cart