ENA321765010 Enamine

1-(2-aminophenyl)-1H-pyrazole-3-carboxamide

  • MDL number MFCD12177643
  • Cas Number : 1153157-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-76440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76440
Availability details for
Quantity
  • Availability
  • Add to Cart