ENA321765014 Enamine

4-chloro-2-methoxy-6-methylpyrimidine

  • MDL number MFCD16987882
  • Cas Number : 97041-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77200
Availability details for
Quantity
  • Availability
  • Add to Cart