ENA321765017 Enamine

4-bromo-N-[2-(methylsulfanyl)ethyl]-1-(propan-2-yl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD12811023
  • Cas Number : 1184272-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76061

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76061
Availability details for
Quantity
  • Availability
  • Add to Cart