ENA321765019 Enamine

6-phenyl-2-sulfanylidene-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD00487109
  • Cas Number : 92126-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77209

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77209
Availability details for
Quantity
  • Availability
  • Add to Cart