ENA321765020 Enamine

2-iodo-5-nitrophenol

  • MDL number MFCD11036284
  • Cas Number : 197243-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76444
Availability details for
Quantity
  • Availability
  • Add to Cart