ENA321765024 Enamine

3-methylthiomorpholine

  • MDL number MFCD11137728
  • Cas Number : 76176-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76445

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76445
Availability details for
Quantity
  • Availability
  • Add to Cart