ENA321765028 Enamine

1-propyl-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD16744015
  • Cas Number : 39213-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77223
Availability details for
Quantity
  • Availability
  • Add to Cart