ENA321765035 Enamine

4-[(5-methylpyridin-2-yl)oxy]aniline

  • MDL number MFCD06825489
  • Cas Number : 229155-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76447
Availability details for
Quantity
  • Availability
  • Add to Cart