ENA321765040 Enamine

[(2,4-dichlorophenyl)(pyridin-3-yl)methyl](methyl)amine

  • MDL number MFCD13328274
  • Cas Number : 1306225-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77250
Availability details for
Quantity
  • Availability
  • Add to Cart