ENA321765042 Enamine

ethyl 2-[(5-nitropyridin-2-yl)amino]acetate

  • MDL number MFCD12157504
  • Cas Number : 98997-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-77253

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77253
Availability details for
Quantity
  • Availability
  • Add to Cart