ENA321765046 Enamine

3-methylpentane-2-sulfonamide

  • MDL number MFCD16672057
  • Cas Number : 1249349-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44...46
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77257

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77257
Availability details for
Quantity
  • Availability
  • Add to Cart