ENA321765052 Enamine

2-(propan-2-yloxy)ethane-1-sulfonamide

  • MDL number MFCD14677086
  • Cas Number : 1249786-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76459

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-76459
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart