ENA321765067 Enamine

1-cyclopentylethan-1-amine

  • MDL number MFCD08704278
  • Cas Number : 38118-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76070

Safety Information

signalword Warning
pcodes P264,P270,P280,P301+P312,P302+P352,P305+P351+P338,P330,P332+P313,P337+P313,P362,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-76070
pictograms GHS07
hstatements H302,H315,H319
Availability details for
Quantity
  • Availability
  • Add to Cart