ENA321765074 Enamine

1-cyano-N,N-dimethylcyclopentane-1-carboxamide

  • MDL number MFCD16104653
  • Cas Number : 1249394-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76476
Availability details for
Quantity
  • Availability
  • Add to Cart