ENA321765077 Enamine

methyl({2-[methyl(2,2,2-trifluoroethyl)amino]ethyl})amine

  • MDL number MFCD12167703
  • Cas Number : 1096348-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76478

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76478
Availability details for
Quantity
  • Availability
  • Add to Cart